Peptone from soybean, enzymatic digest
Synonym(s):
Peptone from soybean
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Special Grade
Showing 1-2 of 2