Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pellicon
®
2 Mini Ultrafiltration Module PLCCC V 0.1 m
2
Synonym(s):
Pellicon
®
2 Mini Ultrafiltration Module
Products
Brand
Millipore (1)
Length (cm)
Length (in)
Quality Segment
MWCO (kDa)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P2C005V01
Pellicon
®
2 Mini Ultrafiltration Module PLCCC V 0.1 m
2
,
filtration area 0.1 m
2
, NMWCO 5 kDa
Expand
View Pricing