PEDOT
Synonym(s):
PEDOT, Poly(2,3-dihydrothieno-1,4-dioxin), PEDOT, tetramethacrylate end-capped, C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG
Linear Formula:
(C6H4O2S)n ·xH2O
Products
Brand
Physical Form
Chemical Composition
Quality Segment
Showing 1-5 of 5
Compare
Product Number
Description
Pricing