![Paliperidone Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/282/543/a962e05e-6317-470e-b8a6-19e643869c3c/640/a962e05e-6317-470e-b8a6-19e643869c3c.jpg)
Paliperidone Related Compound B
![Paliperidone Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/282/543/a962e05e-6317-470e-b8a6-19e643869c3c/640/a962e05e-6317-470e-b8a6-19e643869c3c.jpg)
Synonym(s):
6-Fluoro-3-(piperidin-4-yl)benzoisoxazole hydrochloride, 6-Fluoro-3-(4-piperidinyl)-1,2-benzoisoxazole monohydrochloride
Empirical Formula (Hill Notation):
C12H13FN2O · HCl
CAS Number:
Molecular Weight:
256.70
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing