Skip to Content
Merck
vibrant-m

Oxybutynin Related Compound B

vibrant-m
Synonym(s):
Methyl α-cyclohexyl-alpha-hydroxybenzenacetate, α-Cyclohexylmandelic acid methyl ester, Methyl cyclohexylphenylglycolate
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade