Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ORBO
™
49P (OVS) Supelpak
™
-20
specially treated
Amberlite
™
XAD
®
-2
(20/40), 270/140 mg
Synonym(s):
ORBO
™
Supelpak
™
20 series Tube
Products
Brand
Supelco (1)
Product Line
Agency
Chemical Composition
OD (mm)
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
20350
ORBO
™
49P (OVS) Supelpak
™
-20
specially treated
Amberlite
™
XAD
®
-2
(20/40), 270/140 mg,
RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Expand
View Pricing