Skip to Content
Merck
Oleocanthal phyproof® Reference Substance

Oleocanthal

Oleocanthal phyproof® Reference Substance
Empirical Formula (Hill Notation):
C17H20O5
CAS Number:
Molecular Weight:
304.34

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing