Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
MB120 Mositure Analyzer
Synonym(s):
Ohaus
®
Moisture Analyzer
Products
Product Category
balances (2)
Manufacturer
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
OH30241169
Ohaus
®
MB120 Mositure Analyzer,
Model MB120, weighing capacity 120 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, Schuko plug
Expand
View Pricing
OH30303304
Ohaus
®
MB120 Mositure Analyzer,
Model MB120, weighing capacity 120 g, resolution: 0.001 g, AC/DC input 210 - 2310 V, UK plug
Expand
View Pricing