![NovaSeptum® GO Preloaded System with Accurate Volume Syringe, 9-port sterile; β-irradiated](/deepweb/assets/sigmaaldrich/product/images/277/891/6b489762-4887-4541-8ea5-3e6e674b3827/640/6b489762-4887-4541-8ea5-3e6e674b3827.jpg)
NovaSeptum® GO Preloaded System with Accurate Volume Syringe, 9-port
![NovaSeptum® GO Preloaded System with Accurate Volume Syringe, 9-port sterile; β-irradiated](/deepweb/assets/sigmaaldrich/product/images/277/891/6b489762-4887-4541-8ea5-3e6e674b3827/640/6b489762-4887-4541-8ea5-3e6e674b3827.jpg)
Synonym(s):
NovaSeptum® GO Accurate Volume Syringes
Products
Product Category
Brand
Product Line
Agency
Quality Segment
Application
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing