![Nordoxepin hydrochloride analytical standard, ≥98.0% (TLC), powder](/deepweb/assets/sigmaaldrich/product/structures/263/281/071e3bbe-68ff-46b4-98ba-4b8675a3cb09/640/071e3bbe-68ff-46b4-98ba-4b8675a3cb09.png)
Nordoxepin hydrochloride
![Nordoxepin hydrochloride analytical standard, ≥98.0% (TLC), powder](/deepweb/assets/sigmaaldrich/product/structures/263/281/071e3bbe-68ff-46b4-98ba-4b8675a3cb09/640/071e3bbe-68ff-46b4-98ba-4b8675a3cb09.png)
Synonym(s):
11(6H)-(3-[Methylamino]propylidene)dibenz[b,e]oxepine
Empirical Formula (Hill Notation):
C18H19NO · HCl
CAS Number:
Molecular Weight:
301.81
Products
Brand
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing