Skip to Content
Merck
N,N′-Dibutylthiourea 97%

N,N′-Dibutylthiourea

N,N′-Dibutylthiourea 97%
Synonym(s):
N,N′-Dibutylthiourea
Linear Formula:
(CH3CH2CH2CH2NH)2CS
CAS Number:
Molecular Weight:
188.33

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing