N-Lauroylsarcosine sodium salt solution
Synonym(s):
Sarcosyl, Sarkosyl NL, Sodium N-dodecanoyl-N-methylglycinate, Sodium N-lauroylsarcosinate solution
Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Products
Product Category
Brand
Formula Weight
pH Value
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing