![Nitazoxanide pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/702/913/22574886-e505-447e-9970-4d3a3827cfc5/640/22574886-e505-447e-9970-4d3a3827cfc5.png)
Nitazoxanide
![Nitazoxanide pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/702/913/22574886-e505-447e-9970-4d3a3827cfc5/640/22574886-e505-447e-9970-4d3a3827cfc5.png)
Synonym(s):
NTZ; 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide, Nitazoxanide
Empirical Formula (Hill Notation):
C12H9N3O5S
CAS Number:
Molecular Weight:
307.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3