Nicorandil impurity A
Synonym(s):
2-[(Pyridin-4-ylcarbonyl)amino]ethyl nitrate, p-Nicorandil
Empirical Formula (Hill Notation):
C8H9N3O4
CAS Number:
Molecular Weight:
211.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing