Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel silver alloy, Cu64Ni12Zn24
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF00546016
Nickel silver alloy, Cu64Ni12Zn24,
foil, 100 x 100mm, 0.125mm thickness
Expand
View Pricing
GF28780397
Nickel silver alloy, Cu64Ni12Zn24,
foil, 2m coil, 0.125mm thickness
Expand
View Pricing
GF46783266
Nickel silver alloy, Cu64Ni12Zn24,
foil, 25 x 25mm, 0.125mm thickness
Expand
View Pricing
GF10042603
Nickel silver alloy, Cu64Ni12Zn24,
foil, 0.5m coil, 0.125mm thickness
Expand
View Pricing
GF54961083
Nickel silver alloy, Cu64Ni12Zn24,
foil, 1m coil, 0.125mm thickness
Expand
View Pricing
GF66862378
Nickel silver alloy, Cu64Ni12Zn24,
foil, 200 x 200mm, 0.125mm thickness
Expand
View Pricing
GF61540534
Nickel silver alloy, Cu64Ni12Zn24,
foil, 50 x 50mm, 0.125mm thickness
Expand
View Pricing