Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel silver alloy, Cu62Ni18Zn20
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 65
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
GF31303279
Nickel silver alloy, Cu62Ni18Zn20,
1000mm tube, 1.4mm outside diameter, 0.3mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF70470165
Nickel silver alloy, Cu62Ni18Zn20,
100mm tube, 1.8mm outside diameter, 0.28mm wall thickness, 1.24mm inside diameter, as drawn
Expand
View Pricing
GF40766069
Nickel silver alloy, Cu62Ni18Zn20,
foil, 300 x 300mm, 0.30mm thickness, as rolled
Expand
View Pricing
GF87775374
Nickel silver alloy, Cu62Ni18Zn20,
100mm tube, 1.8mm outside diameter, 0.175mm wall thickness, 1.45mm inside diameter, as drawn
Expand
View Pricing
GF00429229
Nickel silver alloy, Cu62Ni18Zn20,
500mm tube, 1.28mm outside diameter, 0.120mm wall thickness, 1.04mm inside diameter, as drawn
Expand
View Pricing
GF35824901
Nickel silver alloy, Cu62Ni18Zn20,
500mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF66536691
Nickel silver alloy, Cu62Ni18Zn20,
500mm tube, 0.91mm outside diameter, 0.19mm wall thickness, 0.53mm inside diameter, as drawn
Expand
View Pricing
GF82455453
Nickel silver alloy, Cu62Ni18Zn20,
foil, 300 x 300mm, 0.60mm thickness, annealed
Expand
View Pricing
GF73451673
Nickel silver alloy, Cu62Ni18Zn20,
foil, 100 x 100mm, 0.60mm thickness, annealed
Expand
View Pricing
GF94687475
Nickel silver alloy, Cu62Ni18Zn20,
foil, 100 x 100mm, 0.30mm thickness, as rolled
Expand
View Pricing
GF19377120
Nickel silver alloy, Cu62Ni18Zn20,
100mm tube, 1.5mm outside diameter, 0.17mm wall thickness, 1.16mm inside diameter, as drawn
Expand
View Pricing
GF64939012
Nickel silver alloy, Cu62Ni18Zn20,
100mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF06932291
Nickel silver alloy, Cu62Ni18Zn20,
foil, 600 x 600mm, 0.60mm thickness, annealed
Expand
View Pricing
GF09374025
Nickel silver alloy, Cu62Ni18Zn20,
349mm tube, 4.0mm outside diameter, 0.25mm wall thickness, 3.5mm inside diameter, as drawn
Expand
View Pricing
GF42283379
Nickel silver alloy, Cu62Ni18Zn20,
1000mm tube, 3.0mm outside diameter, 0.075mm wall thickness, 2.85mm inside diameter, as drawn
Expand
View Pricing
GF52740536
Nickel silver alloy, Cu62Ni18Zn20,
200mm tube, 1.8mm outside diameter, 0.28mm wall thickness, 1.24mm inside diameter, as drawn
Expand
View Pricing
GF58929957
Nickel silver alloy, Cu62Ni18Zn20,
200mm tube, 1.5mm outside diameter, 0.17mm wall thickness, 1.16mm inside diameter, as drawn
Expand
View Pricing
GF09638411
Nickel silver alloy, Cu62Ni18Zn20,
200mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, as drawn
Expand
View Pricing
GF94447503
Nickel silver alloy, Cu62Ni18Zn20,
1000mm tube, 2.3mm outside diameter, 0.2mm wall thickness, 1.9mm inside diameter, as drawn
Expand
View Pricing
GF63605392
Nickel silver alloy, Cu62Ni18Zn20,
750mm tube, 1.8mm outside diameter, 0.45mm wall thickness, 0.9mm inside diameter, as drawn
Expand
View Pricing
Page 1 of 4
Page 1 of 4