![n-Hexanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/332/274/3d9cb14b-6cff-46be-95ea-4356f969556f/640/3d9cb14b-6cff-46be-95ea-4356f969556f.png)
n-Hexanol
![n-Hexanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/332/274/3d9cb14b-6cff-46be-95ea-4356f969556f/640/3d9cb14b-6cff-46be-95ea-4356f969556f.png)
Synonym(s):
1-Hexanol, Hexyl alcohol
Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing