Skip to Content
Merck
Neurokinin B ≥95% (HPLC)

Neurokinin B

Neurokinin B ≥95% (HPLC)
Synonym(s):
β-Neurokinin, Neuromedin K
Empirical Formula (Hill Notation):
C55H79N13O14S2
CAS Number:
Molecular Weight:
1210.42

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing