Skip to Content
Merck
N-Carbethoxyphthalimide 96%

N-Carbethoxyphthalimide

N-Carbethoxyphthalimide 96%
Synonym(s):
N-Ethoxycarbonylphthalimide, Ethyl 1,3-dioxo-2-isoindolinecarboxylate, Ethyl phthalimide-N-carboxylate
Empirical Formula (Hill Notation):
C11H9NO4
CAS Number:
Molecular Weight:
219.19
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing