Skip to Content
Merck
N-Boc-2,3-dihydro-1H-pyrrole 95%

N-Boc-2,3-dihydro-1H-pyrrole

N-Boc-2,3-dihydro-1H-pyrrole 95%
Synonym(s):
1-(tert-Butoxycarbonyl)-2,3-dihydro-1H-pyrrole, N-Boc-2-pyrroline, tert-Butyl 2,3-dihydro-1H-pyrrole-1-carboxylate
Empirical Formula (Hill Notation):
C9H15NO2
CAS Number:
Molecular Weight:
169.22

Products

Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing