Naproxen
Synonym(s):
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Linear Formula:
CH3OC10H6CH(CH3)CO2H
Products
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing