![N-Acetyl-2-chloro-2-deoxyneuraminic acid methyl ester 4,7,8,9-tetraacetate](/deepweb/assets/sigmaaldrich/product/structures/236/874/57eecd8f-958f-466c-8248-71fd9103b13f/640/57eecd8f-958f-466c-8248-71fd9103b13f.png)
N-Acetyl-2-chloro-2-deoxyneuraminic acid methyl ester 4,7,8,9-tetraacetate
![N-Acetyl-2-chloro-2-deoxyneuraminic acid methyl ester 4,7,8,9-tetraacetate](/deepweb/assets/sigmaaldrich/product/structures/236/874/57eecd8f-958f-466c-8248-71fd9103b13f/640/57eecd8f-958f-466c-8248-71fd9103b13f.png)
Synonym(s):
5-(Acetylamino)-2-chloro-2,3,5-trideoxy-D-glycero-β-D-galacto-2-nonulopyranosonic acid methyl ester 4,7,8,9-tetraacetate, N-Acetyl-2-chloro-2-deoxy-β-neuraminic acid methyl ester 4,7,8,9-tetraacetate, Methyl N-acetyl-4,7,8,9-tetra-O-acetyl-2-chloro-2-deoxy-β-neuraminate
Empirical Formula (Hill Notation):
C20H28ClNO12
CAS Number:
Molecular Weight:
509.89
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing