![Nα-Acetyl-L-ornithine](/deepweb/assets/sigmaaldrich/product/structures/264/631/8c18992c-2596-404f-8266-f5a1143a59cc/640/8c18992c-2596-404f-8266-f5a1143a59cc.png)
Nα-Acetyl-L-ornithine
![Nα-Acetyl-L-ornithine](/deepweb/assets/sigmaaldrich/product/structures/264/631/8c18992c-2596-404f-8266-f5a1143a59cc/640/8c18992c-2596-404f-8266-f5a1143a59cc.png)
Empirical Formula (Hill Notation):
C7H14N2O3
CAS Number:
Molecular Weight:
174.20
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing