![Myricitrin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/146/538/438b6e24-130e-4e3e-a5cc-725fadfe55d1/640/438b6e24-130e-4e3e-a5cc-725fadfe55d1.png)
Myricitrin
![Myricitrin analytical standard](/deepweb/assets/sigmaaldrich/product/structures/146/538/438b6e24-130e-4e3e-a5cc-725fadfe55d1/640/438b6e24-130e-4e3e-a5cc-725fadfe55d1.png)
Synonym(s):
3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside, Myricetin 3-O-α-L-rhamnopyranoside, Myricetin 3-O-rhamnoside, Myricitroside
Empirical Formula (Hill Notation):
C21H20O12
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing