Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
mPEG20K
Synonym(s):
mPEG-MAL, mPEG-Succinimidyl Carboxymethyl Ester
Linear Formula:
(CH
3
O(CH
2
CH
2
O)nCH
2
COOC
4
H
4
O
2
N
Products
Brand
Sigma-Aldrich (4)
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
JKA3115
mPEG20K,
average M
n
20,000, Maleimide, thiol reactive
Expand
View Pricing
JKA3100
mPEG20K,
average M
N
20,000, Silane
Expand
View Pricing
JKA3003
mPEG20K,
average M
n
20,000, Succinimidyl Carboxymethyl Ester, amine reactive
Expand
View Pricing
JKA3048
mPEG20K,
average M
N
20,000, Biotin
Expand
View Pricing