Morphine related compound B
Synonym(s):
Pseudomorphine, 2,2′-Bimorphine, 2,2′-Dehydrodimorphine, Oxydimorphine
Empirical Formula (Hill Notation):
C34H36N2O6
CAS Number:
Molecular Weight:
568.66
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing