Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molecular sieves, 3 Å
Synonym(s):
Molecular Sieve UOP Type 3Å
Linear Formula:
K
n
Na
12-n
[(AlO
2
)
12
(SiO
2
)
12
] · xH
2
O
CAS Number:
308080-99-1
Products
Product Category
sieves (4)
Brand
Sigma-Aldrich (4)
Physical Form
Pore Size (Å)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
208582
Molecular sieves, 3 Å,
beads, 8-12 mesh
Expand
View Pricing
208574
Molecular sieves, 3 Å,
beads, 4-8 mesh
Expand
View Pricing
334286
Molecular sieves, 3 Å,
pellets, ~1.6 mm
Expand
View Pricing
334278
Molecular sieves, 3 Å,
pellets, 3.2 mm
Expand
View Pricing