![Modafinil Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/120/332/2b48ced6-a29e-45e9-8ab2-c73e64c043f5/640/2b48ced6-a29e-45e9-8ab2-c73e64c043f5.png)
Modafinil Related Compound A
![Modafinil Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/120/332/2b48ced6-a29e-45e9-8ab2-c73e64c043f5/640/2b48ced6-a29e-45e9-8ab2-c73e64c043f5.png)
Synonym(s):
2-[(Diphenylmethyl)sulfinyl]acetic acid
Empirical Formula (Hill Notation):
C15H14O3S
CAS Number:
Molecular Weight:
274.33
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing