Mirtazapine Related Compound A
Synonym(s):
Desmethylmirtazapine, 1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, N-Demethylmirtazapine
Empirical Formula (Hill Notation):
C16H17N3
CAS Number:
Molecular Weight:
251.33
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing