![Metyrosine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/745/a7f51350-89fe-47bb-a5f6-d3329d478192/640/a7f51350-89fe-47bb-a5f6-d3329d478192.png)
Metyrosine
![Metyrosine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/745/a7f51350-89fe-47bb-a5f6-d3329d478192/640/a7f51350-89fe-47bb-a5f6-d3329d478192.png)
Synonym(s):
α-Methyl-L-tyrosine, L-2-Methyl-3-(4-hydroxyphenyl)alanine, L-AMPT
Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing