Metronidazole benzoate
Synonym(s):
2-Methyl-5-nitro-1H-imidazole-1-ethyl benzoate
Empirical Formula (Hill Notation):
C13H13N3O4
CAS Number:
Molecular Weight:
275.26
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade