Skip to Content
Merck
Metoprolol tartrate European Pharmacopoeia (EP) Reference Standard

Metoprolol tartrate

Metoprolol tartrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(±)-Metoprolol (+)-tartrate salt, (±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor
Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81

Products

Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique