Skip to Content
Merck
Methylprednisolone acetate ≥98% (HPLC)

Methylprednisolone Acetate

Methylprednisolone acetate ≥98% (HPLC)
Synonym(s):
11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate, (6α,11β)-21-(Acetyloxy)-11,17-dihydroxy-6-methylpregna-1,4-diene-3,20-dione acetate, 11β,17α,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate, 21-Acetoxy-11β,17-dihydroxy-6α-methylpregna-1,4-diene-3,20-dione acetate, 6-Methylprednisolone acetate, Depo-Medro acetatel
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
EC Number:

Products

Brand
Color
Feature
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique