Skip to Content
Merck
Methyl heneicosanoate analytical standard

Methyl heneicosanoate

Methyl heneicosanoate analytical standard
Synonym(s):
Heneicosanoic acid methyl ester
Linear Formula:
CH3(CH2)19COOCH3
CAS Number:
Molecular Weight:
340.58
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing