
Methyl-d3-triphenylphosphonium iodide

Linear Formula:
CD3P(C6H5)3I
CAS Number:
Molecular Weight:
407.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1