
Mesosulfuron-methyl

Empirical Formula (Hill Notation):
C17H21N5O9S2
CAS Number:
Molecular Weight:
503.51
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing