![Meptazinol hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/943/480/14de0325-d2b3-4835-a9cc-6796a4526b0c/640/14de0325-d2b3-4835-a9cc-6796a4526b0c.png)
Meptazinol hydrochloride
![Meptazinol hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/943/480/14de0325-d2b3-4835-a9cc-6796a4526b0c/640/14de0325-d2b3-4835-a9cc-6796a4526b0c.png)
Synonym(s):
3-(3-ethylhexahydro-1-methyl-1H-azepin-3-yl)-phenol hydrochloride, IL-22811 hydrochloride, WY-22811 hydrochloride
Empirical Formula (Hill Notation):
C15H23NO· HCl
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing