Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MAXYMum Recovery
®
ultra micro pipette tips
Products
Product Category
pipette tips (4)
Manufacturer
Color
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
AXYT400L
MAXYMum Recovery
®
ultra micro pipette tips,
for Eppendorf
®
, size 0.5-10 μL, pkg of 20x1000tips/cs
Expand
View Pricing
AXYT400LR
MAXYMum Recovery
®
ultra micro pipette tips,
for Eppendorf
®
racked, size 0.5-10 μL, pkg of 5x960tips/cs
Expand
View Pricing
AXYT400LRS
MAXYMum Recovery
®
ultra micro pipette tips,
for Eppendorf
®
racked, size 0.5-20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYT300LSTKS
MAXYMum Recovery
®
ultra micro pipette tips,
for Pipetman
®
, size 0.5-10 μL, pkg of 10x480tips/cs, sterile
Expand
View Pricing