Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MAXYMum Recovery
®
pipette reference tips
Products
Product Category
pipette tips (5)
Manufacturer
Color
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
AXYTR222CL
MAXYMum Recovery
®
pipette reference tips,
universal fit, size 200 μL, pkg of 20x1000tips/cs
Expand
View Pricing
AXYTR222CLR
MAXYMum Recovery
®
pipette reference tips,
universal fit, racked, size 200 μL, pkg of 5x960tips/cs
Expand
View Pricing
AXYTR222CLRS
MAXYMum Recovery
®
pipette reference tips,
universal fit, racked, size 200 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTR222CLSTKS
MAXYMum Recovery
®
pipette reference tips,
universal fit, stacked rack, size 200 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTR222CLSTK
MAXYMum Recovery
®
pipette reference tips,
universal fit, stacked rack, size 200 μL, pkg of 5x960tips/cs
Expand
View Pricing