Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Manganese-nickel alloy, Mn88Ni12
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 50
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF76999516
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.050mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF73406280
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.025mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF83900820
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF60386991
Manganese-nickel alloy, Mn88Ni12,
foil, 50 x 50mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF75683098
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF63107647
Manganese-nickel alloy, Mn88Ni12,
foil, 10mm disks, 0.050mm thickness, as rolled
Expand
View Pricing
GF50998764
Manganese-nickel alloy, Mn88Ni12,
foil, 4mm disks, 0.050mm thickness, as rolled
Expand
View Pricing
GF98593284
Manganese-nickel alloy, Mn88Ni12,
foil, 50 x 50mm, 0.025mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF79000613
Manganese-nickel alloy, Mn88Ni12,
foil, 50 x 50mm, 0.050mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF45128864
Manganese-nickel alloy, Mn88Ni12,
foil, 50 x 50mm, 0.050mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF38274408
Manganese-nickel alloy, Mn88Ni12,
foil, 15mm disks, 0.25mm thickness, as rolled
Expand
View Pricing
GF29575648
Manganese-nickel alloy, Mn88Ni12,
foil, 8mm disks, 0.125mm thickness, as rolled
Expand
View Pricing
GF68236601
Manganese-nickel alloy, Mn88Ni12,
foil, 6mm disks, 0.125mm thickness, as rolled
Expand
View Pricing
GF89075122
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.038mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF64000673
Manganese-nickel alloy, Mn88Ni12,
foil, 50 x 50mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF74754759
Manganese-nickel alloy, Mn88Ni12,
foil, 25 x 25mm, 0.0125mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF93862974
Manganese-nickel alloy, Mn88Ni12,
foil, 100 x 100mm, 0.0125mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF72306198
Manganese-nickel alloy, Mn88Ni12,
foil, 10mm disks, 0.25mm thickness, as rolled
Expand
View Pricing
GF46815981
Manganese-nickel alloy, Mn88Ni12,
foil, 4mm disks, 0.25mm thickness, as rolled
Expand
View Pricing
GF48374523
Manganese-nickel alloy, Mn88Ni12,
foil, 100 x 100mm, 0.25mm thickness, as rolled
Expand
View Pricing
Page 1 of 3
Page 1 of 3