Skip to Content
Merck
Magnesium Standard for ICP TraceCERT®, 1 g/L Mg in nitric acid (nominal concentration)

Magnesium Standard for ICP

Magnesium Standard for ICP TraceCERT®, 1 g/L Mg in nitric acid (nominal concentration)
Synonym(s):
Magnesium nitrate hexahydrate
Linear Formula:
Mg(NO3)2 · 6 H2O
CAS Number:
Molecular Weight:
256.41

Products

Brand
Formula Weight
Product Line
Format
Element
Quality Segment
Application
Special Grade
Technique