![Lucifer Yellow CH dipotassium salt powder](/deepweb/assets/sigmaaldrich/product/structures/123/615/a5993b7e-79a0-43f8-95a4-58d5d097c672/640/a5993b7e-79a0-43f8-95a4-58d5d097c672.png)
Lucifer Yellow CH dipotassium salt
![Lucifer Yellow CH dipotassium salt powder](/deepweb/assets/sigmaaldrich/product/structures/123/615/a5993b7e-79a0-43f8-95a4-58d5d097c672/640/a5993b7e-79a0-43f8-95a4-58d5d097c672.png)
Empirical Formula (Hill Notation):
C13H9K2N5O9S2
CAS Number:
Molecular Weight:
521.57
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing