![L-tert-Leucine 99%](/deepweb/assets/sigmaaldrich/product/structures/266/646/39725902-0c04-4892-9f9c-564bbe956650/640/39725902-0c04-4892-9f9c-564bbe956650.png)
L-tert-Leucine
![L-tert-Leucine 99%](/deepweb/assets/sigmaaldrich/product/structures/266/646/39725902-0c04-4892-9f9c-564bbe956650/640/39725902-0c04-4892-9f9c-564bbe956650.png)
Synonym(s):
(S)-2-Amino-3,3-dimethylbutyric acid, L-α-tert-Butylglycine
Linear Formula:
(CH3)3CCH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing