Skip to Content
Merck
L-Phenyl-13C6-alanine API for Clinical Studies, ≥99 atom % 13C

L-Phenyl-13C6-alanine

L-Phenyl-13C6-alanine API for Clinical Studies, ≥99 atom % 13C
Linear Formula:
13C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
171.15

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing