Lomustine
Synonym(s):
1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea, Lomustine
Empirical Formula (Hill Notation):
C9H16ClN3O2
CAS Number:
Molecular Weight:
233.70
Products
Brand
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade