![Lithium thiophenolate solution 1.0 M in THF](/deepweb/assets/sigmaaldrich/product/structures/114/885/7212d45f-346a-4e79-8975-340ff21c5584/640/7212d45f-346a-4e79-8975-340ff21c5584.png)
Lithium thiophenolate solution
![Lithium thiophenolate solution 1.0 M in THF](/deepweb/assets/sigmaaldrich/product/structures/114/885/7212d45f-346a-4e79-8975-340ff21c5584/640/7212d45f-346a-4e79-8975-340ff21c5584.png)
Synonym(s):
Thiophenol lithium salt
Linear Formula:
C6H5SLi
CAS Number:
Molecular Weight:
116.11
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing