Skip to Content
Merck
Lithium nitrate battery grade, anhydrous, ≥99.9% trace metals basis

Lithium nitrate

Lithium nitrate battery grade, anhydrous, ≥99.9% trace metals basis
Synonym(s):
Lithium nitrate
Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group