Skip to Content
Merck
Levocetirizine Dihydrochloride pharmaceutical secondary standard, certified reference material

Levocetirizine dihydrochloride

Levocetirizine Dihydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
Levocetirizine dihydrochloride, (-)-Cetirizine dihydrochloride; 2-[2-[4-[(R)-(4-chlorophenyl)-phenyl-methyl]piperazin-1-yl]ethoxy]acetic acid dihydrochloride
Empirical Formula (Hill Notation):
C21H25ClN2O3·2HCl
CAS Number:
Molecular Weight:
461.81

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3