
Leu-Ala hydrate

Synonym(s):
L-Leucyl-L-alanine hydrate
Linear Formula:
(CH3)2CHCH2CH(NH2)CONHCH(CH3)COOH · xH2O
CAS Number:
Molecular Weight:
202.25 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1