Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
LB Broth
Products
Product Category
media (2)
Brand
Millipore (2)
Color
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.10285
LB Broth,
GranuCult
®
plus, granular, pack of 500 g or 5 kg, for
Escherichia coli
Expand
View Pricing
51208
LB Broth,
NutriSelect
®
Plus, High salt, powder, pack of 500 g or 2.5 kg, non-sterile, nonselective for
Escherichia coli
Expand
View Pricing